Recombinant Human CXCL11/I-TAC Protein 1 * 100 µG
Catalog # ABCAAB283901-100
Supplier: Abcam
Frequently Bought Together
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Deep well plates, 24-well, Axygen®
Supplier: Corning
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
VWR®, Square Bottles, PET
Supplier: LAB3 OWN PRODUCTION
Storage Boxes
Supplier: EPPENDORF
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Storage canes for cryotubes CryoLine™, Nunc™
Supplier: Thermo Fisher Scientific
Multiple well cell culture plates, Costar®
Supplier: Corning
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, Low-Binding
Supplier: VWR Collection
Scoops for foodstuffs, SteriPlast®
Supplier: BURKLE
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
Supplier: LAB3 OWN PRODUCTION
VWR® Cryoboxes, Double Size, 133x257 mm
Supplier: VWR Collection