ADENINE (8-13C, 95%) MAY CONTAIN OP TO 7% 2-13C 1 * 500 mg
CAMBCLM-1654-0.5
ADENINE (8-13C, 95%) MAY CONTAIN OP TO 7% 2-13C 1 * 500 mg
CAMBCLM-1654-0.5
Frequently Bought Together
Sodium chloride ≥99%, GPR RECTAPUR®
: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
: VWR Chemicals
Ammonium chloride ≥99%, GPR RECTAPUR®
: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals