FORMAMIDE (D3 98%) 1 * 5 g
Catalog # CAMBDLM-1864-5
Supplier: CAMBRIDGE ISOTOPE LABS
Frequently Bought Together
N-HEPTANE FOR LIQUID CHROMATOGRAPHY 1 * 185 L
Supplier: Merck
Methanol ≥99.9% ACS, Spectrophotometric Grade for spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Cyclohexane for pesticide residue analysis
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
n-Hexane, anhydrous ≥96.0% (by GC)
Supplier: TCI
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
Amyl Alcohol according to NF V 04-210 for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem