Order Entry
Denmark
ContactUsLinkComponent
Microscope slides, 76x52 mm, thickness 1.35 mm +/- 0.15 mm, white glass, edges ground at 90, surface frosted completely, in boxes of 50 1 * 1.000 items
Catalog # MARI1123720
Supplier:  MARIENFELD
Microscope slides, 76x52 mm, thickness 1.35 mm +/- 0.15 mm, white glass, edges ground at 90, surface frosted completely, in boxes of 50 1 * 1.000 items
Catalog # MARI1123720
Supplier:  MARIENFELD
Frequently Bought Together