VWR® Buffer Solutions, Secondary Standards
7.000, ±0,010, 25 °C, Plastic bottle, 500 mL
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
- Accurate to ±0,010 pH units (except buffer 12,454 with accuracy ±0,050)
- Solution standardised at 25 °C
- Fully traceable for optimum pH calibration (traceable to NIST, DIN 19266 and IUPAC)
- Temperature dependence data is printed on the labels as are batch number and expiry date
Documents
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Reference temperature:25 °C
- pH accuracy:±0,010
- pH value:7.000
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



