Order Entry
Denmark
Orders LinkContactUsLinkComponent
Bottle Holders

Bottle Holders

Bottle holder, 1 bottle wall mounted, 110×110×120 mm
Catalog # TEKMHWBH-0500-1
Supplier:   TEKNOMEK
Bottle Holders
Catalog # TEKMHWBH-0500-1
Supplier:  TEKNOMEK
Bottle Holders
Bottle Holders

Bottle holder, 1 bottle wall mounted, 110×110×120 mm

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

Our bottle holders come in a range of sizes.

  • Brushed dull polish finish
  • Easy clean design
  • 80 mm diameter hole

Our bottle holders are the ideal solution to your storage needs, coming in a range of sizes (1, 2 or 3 bottle capacities) to suit your requirements.

The bottle holders are manufactured in fully welded 304 grade stainless steel with a brushed dull polish finish.

The stainless steel bottle holder is suitable for housing 500 ml bottles with 80 mm diameter rings, ensuring they can hold a wide variety of bottles.

Specifications
  • Description:
    Bottle holder, 1 bottle wall mounted
  • Seq:
    01
  • W×D×H:
    110×110×120 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.