Order Entry
Denmark
ContactUsLinkComponent
Objective N PLAN L 40x/0.55 CORR Free working distance: 3.3 - 1.9 mm For use with cover glass of 0-2 mm Correction collar Useful and recommended for Integrated Leica Modulation Contrast (IMC) 1 * 1 items
Catalog # LEIM11506297
Supplier:  LEICA MICROSYSTEMS
Objective N PLAN L 40x/0.55 CORR Free working distance: 3.3 - 1.9 mm For use with cover glass of 0-2 mm Correction collar Useful and recommended for Integrated Leica Modulation Contrast (IMC) 1 * 1 items
Catalog # LEIM11506297
Supplier:  LEICA MICROSYSTEMS
Frequently Bought Together