ETHANOL (2-13C, 99%) (<6% H2O) 1 * 0,5 g
Catalog # CAMBCLM-130-0.5
Leverandør: CAMBRIDGE ISOTOPE LABS
ETHANOL (2-13C, 99%) (<6% H2O) 1 * 0,5 g
Catalog # CAMBCLM-130-0.5
Leverandør: CAMBRIDGE ISOTOPE LABS
Frequently Bought Together
Ethanol, GPR RECTAPUR®
Leverandør: VWR Chemicals
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Leverandør: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverandør: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Leverandør: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverandør: VWR Chemicals
Water, GPR RECTAPUR®
Leverandør: VWR Chemicals
Acetone ≥99%, TECHNICAL
Leverandør: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverandør: VWR Chemicals
Chloroform, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Leverandør: Merck
2-Propanol ≥98%, TECHNICAL
Leverandør: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Leverandør: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverandør: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Leverandør: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverandør: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverandør: VWR Chemicals