Pulsafeeder Solenoid Metering Pump Five-Function Valve, PVDF, 100 psi, 1/2" OD
Catalog # MFLX74113-88
Supplier: Avantor Fluid Handling
Specifications
See all product configurations- Warranty:
- 1 year
- 3 Months
- 3 Months
- 1 year
- Currently Viewing
Specifications
About this item
Optimize performance of your Pulsafeeder Solenoid Metering Pump.
- Choose from a wide range of replacement parts and accessories
Complete and customize a pump system for any application with a variety of valves and mounting options. Service kits are also available to make routine maintenance easy.
- Description:PVDF five-function valve, compression fitting, 1/2" tubing connectors, 100 psi max
- Environmentally Preferable:
- Warranty:1 year
- Pk:1
Frequently Bought Together