Ordreindtastning
Denmark
ContactUsLinkComponent
Fitting, Brass, Straight, Compression Bulkhead Union, 1/4" OD
Fitting, Brass, Straight, Compression Bulkhead Union, 1/4" OD
Catalog # MFLX31412-13
undefined
Fitting, Brass, Straight, Compression Bulkhead Union, 1/4" OD
Catalog # MFLX31412-13
About this item
Get a secure seal at higher pressures.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rates to 220 psi at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).
  • Tilslutningstype:
    Compression Union
  • Flow pattern:
    Straight
  • Beskrivelse:
    Fitting, Brass, Straight, Compression Bulkhead Union, ¼" OD
  • Environmentally Preferable:
  • Garanti:
    LTD life time
  • Pakn:
    1
  • Fitting material:
    Brass
Frequently Bought Together