HYDROCHLORIC ACID 2.0N SOLUTION 1 * 25 L
Catalog # 0358.9025
Supplier: Avantor
Frequently Bought Together
Triethylene glycol dimethyl ether stabilised, Sigma-Aldrich®
Supplier: Merck
n-Pentane ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
N-HEPTANE FOR LIQUID CHROMATOGRAPHY 1 * 185 L
Supplier: Merck
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
Diisopropyl ether 98+% stabilised, pure
Supplier: Thermo Fisher Scientific
n-Heptane 99+%, pure
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Pentane, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific
Chloroform 99.8% stabilised for spectroscopy
Supplier: Thermo Fisher Scientific
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific