Order Entry
Denmark
ContactUsLinkComponent
SEATING BENCH 2 SHELF 1000 X 350 Dimensions 1000 x 350 x 450mm (LxWxH) Spacing between the shelves 130mm Floor Clearance: 48mm Payload: 250Kg 304 Grade Stainless steel 4 x Stainless steel non marking leveling feet 1 * 1 items
  TEKMCRSB-S210
 :  TEKNOMEK
undefined
SEATING BENCH 2 SHELF 1000 X 350 Dimensions 1000 x 350 x 450mm (LxWxH) Spacing between the shelves 130mm Floor Clearance: 48mm Payload: 250Kg 304 Grade Stainless steel 4 x Stainless steel non marking leveling feet 1 * 1 items
  TEKMCRSB-S210
 :  TEKNOMEK
Frequently Bought Together