UDP--D-GALACTOSE (SODIUM SALT) 1 * 5 mg
Catalog # CAYM34664-5
Supplier: Cayman Chemical
Frequently Bought Together
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Mineral oil heavy, white stabilised USP, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
n-Hexane ≥99%
Supplier: Thermo Fisher Scientific
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Glycerine (synthetic) 99+%, pure
Supplier: Thermo Fisher Scientific
80 Octane Blend, PRF B&J Brand, meets requirements of ASTM D2699 and D2700, Burdick & Jackson™
Supplier: Honeywell Chemicals
Toluene ≥99.5% ACS
Supplier: Thermo Fisher Scientific
ACETONE PURE PHARMA GRADE 1 * 200 L
Supplier: PanReac AppliChem
Petroleum spirit, 35…60 °C ACS
Supplier: Thermo Fisher Scientific
Trifluoroacetic acid 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Acetone (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 1 L
Supplier: PanReac AppliChem
Water, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Diethyl ether ≥99.5% (by GC) stabilised, Puriss., meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals