Cytokeratin 10 (C-Term) Rabbit Monoclonal Antibody 1 * 100 µl
CAYM32313-100
Frequently Bought Together
Reagent/centrifuge tube with screw cap
: Greiner Bio-One
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR® Premium Line, Serological Pipettes
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection