Order Entry
Denmark
Orders LinkContactUsLinkComponent
Coupling body plugs
Coupling body plugs
Coupling body plug, with class VI silicone O-ring, MPX series, Polysulfone
Catalog # CPCCMPX30039MB25
Supplier:   CPC COLDER
Coupling body plugs
Catalog # CPCCMPX30039MB25
Supplier:  CPC COLDER
Coupling body plugs
Coupling body plug, with class VI silicone O-ring, MPX series, Polysulfone
Price
3.490,00 kr
Log in for availability
1 * 25 Items
Product Details & Documents
Class VI coupling body plug with Class VI silicone O-ring.
Specifications
  • Description:
    Coupling body plug, with class VI silicone O-ring, MPX series
  • Material:
    Polysulfone
  • Seq:
    04