Order Entry
Denmark
Orders LinkContactUsLinkComponent
MPX series ¹/₂" flow, quick disconnect coupling for bio and pharma
MPX series ¹/₂" flow, quick disconnect coupling for bio and pharma
¹/₂" Hose barb non valved insert (male connector), polysulphone
Katalog # 212-0837
Leverandør:   CPC COLDER
MPX series ¹/₂" flow, quick disconnect coupling for bio and pharma
Katalog # 212-0837
Leverandør:  CPC COLDER
MPX series ¹/₂" flow, quick disconnect coupling for bio and pharma
MPX series ¹/₂" flow, quick disconnect coupling for bio and pharma
¹/₂" Hose barb non valved insert (male connector), polysulphone
Price
22.850,00 kr
Log ind for tilgængelighed
1 * 250 Items
Product Details & Documents
These ¹/₂" flow quick disconnect couplings are manufactured specifically for the bio and pharma markets.
Materials of construction include animal free (BSE/TSE) USP class 6 polysulphone and polycarbonate, Buna - n and platinum cured USP class 6 silicone oring seals.
USP Class 6 and animal free (BSE/TSE) materials are appropriate for the Bio, Pharma and Life Sciences with single use dispoability in mind.
 : USP Class VI, BSE/TSE free materials (ADCF)
 : Accessories not included
Specifikationer
  • Description:
    ¹/₂" Hose barb non valved insert (male connector), polysulphone
  • Seq:
    08
Anbefalinger