Order Entry
Denmark
Orders LinkContactUsLinkComponent
Clipboards
Clipboards
Clipboard, A3 landscape with backclip, 350×470 mm
Catalog # TEKMWW0205LSSCCLIP
Supplier:   TEKNOMEK
Clipboards
Catalog # TEKMWW0205LSSCCLIP
Supplier:  TEKNOMEK
Clipboards
Clipboard, A3 landscape with backclip, 350×470 mm
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
The clipboards come in a range of options.
  • Brushed dull polish finish
  • Spring loaded stainless steel clip
  • Available in portrait or landscape
  • Material: Fully welded 304-grade stainless steel
The stainless steel clipboards come in sizes A4 and A3, and can be supplied with or without a clip at the back to hang them from our hanging rails.
Specifications
  • Description:
    Clipboard, A3 landscape with backclip
  • Seq:
    06
  • W×H:
    350×470 mm