Clipboards
Clipboard, A4 landscape with backclip, 240×400 mm
Catalog # TEKMWW0200SSCCLIPL
Supplier: TEKNOMEK
Jump to:
Product Details & Documents
The clipboards come in a range of options.
- Brushed dull polish finish
- Spring loaded stainless steel clip
- Available in portrait or landscape
- Material: Fully welded 304-grade stainless steel
The stainless steel clipboards come in sizes A4 and A3, and can be supplied with or without a clip at the back to hang them from our hanging rails.
Specifications
- Description:Clipboard, A4 landscape with backclip
- Seq:02
- W×H:240×400 mm