Blackout paper labels
Blackout paper labels, Paper, White, 101,6×25, 4 mm
Catalog # GAINBOP-127C1-1WH
Supplier: GA
Jump to:
Product Details & Documents
These cover-up labels are available in roll format, and are compatible with all standard thermal transfer barcode printers.
- Unique opaque design allows the labels to block out all pre-existing information
- For general office, laboratory, and warehouse applications
- Compatible with all major thermal transfer printer brands
Permanent blackout paper labels feature a unique opaque design that allows them to obscure any existing printed information including serial numbers, barcodes, and text. Perfectly suited for general use office, laboratory, and warehouse applications.
Specifications
- Colour:White
- Description:Blackout paper labels
- Dimensions:101,6×25, 4 mm
- Material:Paper
- Seq:01