Order Entry
Denmark
ContactUsLinkComponent
2% Gel Cups (TAE) w/ GelGreen DNA Stain: includes: Ten GelCups with preweighed 2% Agarose and GelGreen DNA Stain in 0.5XTAEbuffer, 100 mL Tris-Acetate-EDTA (TAE) buffer concentrate 1 * 10 items
Catalog # EMBIM3203TAE
Supplier:  EMBITEC
2% Gel Cups (TAE) w/ GelGreen DNA Stain: includes: Ten GelCups with preweighed 2% Agarose and GelGreen DNA Stain in 0.5XTAEbuffer, 100 mL Tris-Acetate-EDTA (TAE) buffer concentrate 1 * 10 items
Catalog # EMBIM3203TAE
Supplier:  EMBITEC
Frequently Bought Together