VWR®, Storage Boxes for Sample Vials and Bottles, PP
Violet, Box with 16 cavities for 20 ml EPA vials, 133×133×70-80 mm
Catalog # 548-1512A
Supplier: VWR Collection
Jump to:
Product Details & Documents
A broad variety of PP storage boxes to securely hold a wide range of vial sizes and bottles. Perfect for space-saving storage in the laboratory or in fridges.
- Transparent lid
- Chemically resistant
- Stackable and robust
- Temperature resistant from –80 to 100 °C
- Easy sample identification (alphanumeric coding) for boxes with 81 cavities
Transperent lids prevents condensation on the closures and thus avoid contamination in the refrigerator.
Specifications
- Colour:Violet
- Description:Box with 16 cavities for 20 ml EPA vials
- Seq:14
- W×D×H:133×133×70-80 mm



