Watch glasses
Watch glass, glass, with fused rim, 180 mm
Catalog # HECH40010018
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Evaporating dishes, made of glass, with fused rim.
- Good chemical restistance
- Available with or without hole
Certifications: DIN 12 341
Specifications
- Ø:180 mm
- Description:Watch glass, glass, with fused rim
- Seq:21
Anbefalinger