Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Short-term storage solutions, 2-8CELLsius

Solution in processing bag, 1.000 mL
  PRPHPP338-1000B
Short-term storage solutions, 2-8CELLsius
  PRPHPP338-1000B
 :  
Short-term storage solutions, 2-8CELLsius
Short-term storage solutions, 2-8CELLsius

Solution in processing bag, 1.000 mL

 :  
 
 
   
1 * 1.000 mL
 

Ready to use, balanced, cytoprotective, protein-free solution with bio-stabilisers for short-term storage, regenerative medicine, stem cell processing and transportation of biological material. Short-term storage can be achieved at 2 to 8 °C or at ambient temperature for 24 to 48 hours. Does not contain any growth factors, hormones, or antibiotics. 2-8CELLsius™ can also be utilised as a cell washing solution.

  • TYPE II drug master file
  • USP grade components/highest grade
  • Supports high cell viability
  • No animal or human proteins
  • No growth factors, hormones or antibiotics
  • Ready to use/sterile membrane filtered

 : cGMP manufactured.

 

 

  • Pk:
    1.000 mL
  • Description:
    Solution in processing bag
  • Cat. No.:
    PRPHPP338-1000B
  • Storage temperature:
    2...8 °C
  • Shelf life:
    20 months from DOM
  • Endotoxin:
    <0,5 EU/ml at 1X