Cryopreservation solutions, defined, 2-8CELLsius + DMSO
DMSO percentage 20%, processing bag, 1.000 mL
Catalog # PRPH338DM20-1000B
Supplier: PROTIDE PHARMACEUTICALS
Cryopreservation solutions, defined, 2-8CELLsius + DMSO
Catalog # PRPH338DM20-1000B
Supplier: PROTIDE PHARMACEUTICALS
CAS Number:
Jump to:
Product Details & Documents
When combined with DMSO, 2-8CELLsius™ can be utilised as a cryopreservation solution. 2-8CELLsius™ + DMSO is a cytoprotective, protein-free solution, with bio-stabilisers that support high post thaw cell viability. Optimal recovery is achieved through a novel approach to cell physiology at ultra-low temperatures.
- TYPE II drug master file
- USP grade components/highest grade
- Supports high cell viability
- No animal or human proteins
- No growth factors, hormones, antibiotics
- Ready to use/sterile membrane filtered
Certifications: cGMP manufactured.
Specifications
Specifications
- Pk:1.000 mL
- Description:DMSO percentage 20%, processing bag
- Cat. No.:PRPH338DM20-1000B
- Storage temperature:2...8 °C
- Shelf life:20 months from DOM
- Endotoxin:<0,5 EU/ml at 1X
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



