Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Cryopreservation solutions, defined, 2-8CELLsius + DMSO

DMSO percentage 10%, glass vial, 50 mL
  PRPHPP338DM10-50
Cryopreservation solutions, defined, 2-8CELLsius + DMSO
  PRPHPP338DM10-50
 :  
Cryopreservation solutions, defined, 2-8CELLsius + DMSO
Cryopreservation solutions, defined, 2-8CELLsius + DMSO

DMSO percentage 10%, glass vial, 50 mL

 :  
 
 
   
1 * 50 mL
 

When combined with DMSO, 2-8CELLsius™ can be utilised as a cryopreservation solution. 2-8CELLsius™ + DMSO is a cytoprotective, protein-free solution, with bio-stabilisers that support high post thaw cell viability. Optimal recovery is achieved through a novel approach to cell physiology at ultra-low temperatures.

  • TYPE II drug master file
  • USP grade components/highest grade
  • Supports high cell viability
  • No animal or human proteins
  • No growth factors, hormones, antibiotics
  • Ready to use/sterile membrane filtered

 : cGMP manufactured.

 

 

  • Pk:
    50 mL
  • Description:
    DMSO percentage 10%, glass vial
  • Cat. No.:
    PRPHPP338DM10-50
  • Storage temperature:
    2...8 °C
  • Shelf life:
    20 months from DOM
  • Endotoxin:
    <0,5 EU/ml at 1X