TCH defined serum-free growth medium
TCH defined serum-free growth medium, 40 mL
Katalog # PRPH3001
Leverandør: PROTIDE PHARMACEUTICALS
TCH defined serum-free growth medium
Katalog # PRPH3001
Leverandør: PROTIDE PHARMACEUTICALS
CAS nummer:
Gå til:
Product Details & Documents
Serum replacement used primarily for human cells and the production of cell secreted proteins. TCH™ is also effective in culturing other mammalian cells without the use of animal proteins. Contains only human proteins (i.e., human serum albumin, human transferrin, recombinant insulin), which are heat treated.
- Defined, serum-free, xeno-free
- No hormones, antibiotics, growth factors, phenol red, Ig
- 50X concentrate
- Can be utilised with a wide variety of basal media
: cGMP manufactured.
Specifikationer
Specifikationer
- Pk:40 mL
- Description:TCH defined serum-free growth medium
- Cat. No.:PRPH3001
- Total protein:570 µg/ml at 1X
- Storage temperature:2...8 °C
- Shelf life:18 months from DOM
- Species:Human
- Endotoxin:<0,5 EU/ml at 1X



