Order Entry
Denmark
ContactUsLinkComponent
SARS-CoV-2 Spike Trimer, His Tag (SPN-C52He) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession QHD43416.1). The mutations (T19R, G142D, EF156-157del, R158G, L452R, T478K, D614G, P681R, D950N) were identified in b 1 * 50 µG
Catalog # ACRBSPN-C52HE-50UG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 Spike Trimer, His Tag (SPN-C52He) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession QHD43416.1). The mutations (T19R, G142D, EF156-157del, R158G, L452R, T478K, D614G, P681R, D950N) were identified in b 1 * 50 µG
Catalog # ACRBSPN-C52HE-50UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together