Order Entry
Denmark
ContactUsLinkComponent
Biotinylated SARS-CoV-2 Spike S1, His,Avitag (S1N-C82E5) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession QHD43416.1). The mutations (T19R, G142D, EF156-157del, R158G, L452R, T478K, D614G, P681R) were identifie o 1 * 200 µG
Catalog # ACRBS1N-C82E5200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated SARS-CoV-2 Spike S1, His,Avitag (S1N-C82E5) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession QHD43416.1). The mutations (T19R, G142D, EF156-157del, R158G, L452R, T478K, D614G, P681R) were identifie o 1 * 200 µG
Catalog # ACRBS1N-C82E5200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together