Order Entry
Denmark
ContactUsLinkComponent
SARS-CoV-2 Spike RBD, His Tag (SPD-C5227) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession Q HD43416.1 (L452Q, F490S)). Mutations L452Q, F490S are identified in the SARS-CoV-2 Lambda Variant (C.37). 1 * 100 µG
Catalog # ACRBSPD-C5227100UG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 Spike RBD, His Tag (SPD-C5227) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession Q HD43416.1 (L452Q, F490S)). Mutations L452Q, F490S are identified in the SARS-CoV-2 Lambda Variant (C.37). 1 * 100 µG
Catalog # ACRBSPD-C5227100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together