Order Entry
Denmark
ContactUsLinkComponent
RSA-Pro X Inserts, in Racks.Includes 96, Clear, 350ul Glass Low Volume Precision Inserts with wide Conical Points in plastic racks. U-2D Brand. 6 Racks / PK. MOST POPULAR 1 * 6 items
Catalog # MICS95296-350-P-PK
Supplier:  MICROSOLV
RSA-Pro X Inserts, in Racks.Includes 96, Clear, 350ul Glass Low Volume Precision Inserts with wide Conical Points in plastic racks. U-2D Brand. 6 Racks / PK. MOST POPULAR 1 * 6 items
Catalog # MICS95296-350-P-PK
Supplier:  MICROSOLV
Frequently Bought Together