Order Entry
Denmark
ContactUsLinkComponent
absorbent, supple, colorfast, wash- and boil-proof, lint-free, width 21 cm, color blue, length 25 cm, material 100 % cellulose 1 * 1 items
Catalog # KAMM1323721
Supplier:  KAMMERER
absorbent, supple, colorfast, wash- and boil-proof, lint-free, width 21 cm, color blue, length 25 cm, material 100 % cellulose 1 * 1 items
Catalog # KAMM1323721
Supplier:  KAMMERER
Frequently Bought Together