Quercetin, anhydrous
Bottle, 1 g
Catalog # APOSBIK9011-1G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 3-5-7-3-4-Pentahydroxyflavone, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-1-benzopyran-4-one, 3,5,7,3',4'-Pentahydroxyflavone
- CAS Number:117-39-5
- Formula:C₁₅H₁₀O₇
- MDL Number:MFCD00149487
- Molecular Weight:302.25 g/mol
- Storage Temperature:Ambient
- EINECS:204-187-1
- UN:2811
- ADR:6.1,III
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Pk:1 g



