KS184 - QA3 - QAC HR TITRANT 1 * 65 mL
Catalog # LOVI56L018465
Supplier: Lovibond
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
Pyridine ≥99%, TECHNICAL
Supplier: VWR Chemicals
Nitric acid 1 mol/l (1 N), AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
(±)-2-Butanol, EMSURE® for analysis, Supelco®
Supplier: Merck
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
p-Xylene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Heptane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Titrants and reagents for volumetric Karl Fischer titrations, HYDRANAL™, Fluka™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals