1H,1H,2H,2H-Perfluorooctane Sulfonic Acid 100 µg/Ml In Methanol 1 * 1 mL
EHERA15987125ME
1H,1H,2H,2H-Perfluorooctane Sulfonic Acid 100 µg/Ml In Methanol 1 * 1 mL
EHERA15987125ME
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection