Condensers for REVOLVE 3M microscope
Turret condenser, long working distance universal, WD 27 mm, NA 0,55 aperture (five position rotating turret includes PHL, PH1, PH2, PH3 phase annuli)
Catalog # 630-3335
Supplier: DISCOVER ECHO
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Turret condenser, long working distance universal, WD 27 mm, NA 0,55 aperture (five position rotating turret includes PHL, PH1, PH2, PH3 phase annuli)
- Seq:02