Order Entry
Denmark
ContactUsLinkComponent
Roller bottle cell culture Cellmaster polystyrene 122/500mm 4250cm 4640ml 5XL long form ribbed surface clear TC standard screw cap sterile 1 * 24 items
Catalog # GREI682672_24
Supplier:  Greiner Bio-One
Roller bottle cell culture Cellmaster polystyrene 122/500mm 4250cm 4640ml 5XL long form ribbed surface clear TC standard screw cap sterile 1 * 24 items
Catalog # GREI682672_24
Supplier:  Greiner Bio-One
Frequently Bought Together