Order Entry
Denmark
Orders LinkContactUsLinkComponent
Funnels with level control, b.safe
Funnels with level control, b.safe
Funnels 180, static dissipative, with level indicator, grounding cable and vertical connector, S60
Catalog # 590-0381
Supplier:   Bohlender
Funnels with level control, b.safe
Catalog # 590-0381
Supplier:  Bohlender
Funnels with level control, b.safe
Funnels 180, static dissipative, with level indicator, grounding cable and vertical connector, S60
Price
2.850,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
b.safe funnels with level control are made of electrically conductive dissipative PE-EX and suitable for use with flammable liquids. Electrically conductive waste canisters are usually black and therefore the filling volume is not visible. This is where the funnel with fill level control comes in.
  • Prevent the waste canisters from overflowing
  • Available for canisters and barrels
  • With lid or splash protector
  • 130 or 180 mm funnel diameter
The funnels are available in two sizes, each with a hinged lid or splash guard. The unique design with slanted thread connection allows the funnel to be aligned horizontally on canisters with slanted necks, without the need for an additional adapter.
The floater is designed to function reliably even with low solvent densities. An earthing cable is included in the scope of delivery and must be connected accordingly to prevent ignition hazards due to electrostatic charging. The integrated sieve retains magnetic stirring bars and can be easily removed for cleaning.
Documents
Specifications
  • Thread:
    S60
  • Description:
    Funnels 180, static dissipative, with level indicator, grounding cable and vertical connector
  • Seq:
    04