Sodium L(+)-ascorbate 98%
Bottle, 500 g
Catalog # APOSOR922808-500G
Supplier: Apollo Scientific
Sodium L(+)-ascorbate 98%
Catalog # APOSOR922808-500G
Supplier: Apollo Scientific
CAS Number: 134-03-2
Jump to:
Product Details & Documents
Synonyms: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
- CAS Number:134-03-2
- Formula:C₆H₇NaO₆
- MDL Number:MFCD00082340
- Molecular Weight:198.11 g/mol
- Melting Pt:219…221 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



