Order Entry
Denmark
Orders LinkContactUsLinkComponent
Valeric acid 99%

Valeric acid 99%

Bottle, 100 g
Catalog # APOSOR55656-100G
Corrosive
Irritant
Supplier:   Apollo Scientific
Valeric acid 99%
Catalog # APOSOR55656-100G
Supplier:  Apollo Scientific
CAS Number:  109-52-4
Valeric acid 99%

Bottle, 100 g

CAS Number:  109-52-4
Price
439,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Pentanoic acid
  • CAS Number:109-52-4
  • Formula:C₅H₁₀O₂
  • MDL Number:MFCD00004413
  • Molecular Weight:102.13 g/mol
  • Boiling Pt:184…186 °C
  • Melting Pt:–34 °C
  • Density:0.938 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • UN:3265
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.