Order Entry
Denmark
Orders LinkContactUsLinkComponent
Valeric acid 99%
Bottle, 25 g
  APOSOR55656-25G
Valeric acid 99%
  APOSOR55656-25G
 :  109-52-4
Valeric acid 99%
Bottle, 25 g
 :  109-52-4
165,00 kr
1 * 25 g
 :  Pentanoic acid
  •  :109-52-4
  •  :C₅H₁₀O₂
  •  :MFCD00004413
  •  :102.13 g/mol
  •  :184…186 °C
  •  :–34 °C
  •  :0.938 g/cm³ (25 °C)
  •  :Ambient
  •  :3265
  •  :8,III
  • Pack type:
    Bottle
  • Pk:
    25 g