Order Entry
Denmark
Orders LinkContactUsLinkComponent
4-(Methoxymethyl)phenol 95%

4-(Methoxymethyl)phenol 95%

Bottle, 1 g
Catalog # APOSOR928265-1G
Supplier:   Apollo Scientific
4-(Methoxymethyl)phenol 95%
Catalog # APOSOR928265-1G
Supplier:  Apollo Scientific
CAS Number:  5355-17-9
4-(Methoxymethyl)phenol 95%

Bottle, 1 g

CAS Number:  5355-17-9
Price
1.740,00 kr
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:5355-17-9
  • Formula:C₈H₁₀O₂
  • MDL Number:MFCD00020186
  • Molecular Weight:138.16 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications