Phenol solution 5% (w/v)
Plastic, 4 L
Catalog # SPCMP-259-4LTPL
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Phenol, 5 Percent (w/v) Solution - Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene
- CAS Number:108-95-2
- Formula:C₆H₅OH
- MDL Number:MFCD00002143
- Molecular Weight:94.11 g/mol
Specifications
Specification Test Results
- Assay (w/v):4.9-5.1 %
Specifications
- Pack type:Plastic
- Pk:4 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



