Order Entry
Denmark
ContactUsLinkComponent
NOTE: THIS FILTER KIT IS NOT FOR USE WITH OXYGEN. COMPATIBLE WITH ICP-MS INSTRUMENTS SUCH AS THE NEXION 1000/2000/350/300. 1 * 1 items
Catalog # PERCN8146004
Supplier:  PerkinElmer
NOTE: THIS FILTER KIT IS NOT FOR USE WITH OXYGEN. COMPATIBLE WITH ICP-MS INSTRUMENTS SUCH AS THE NEXION 1000/2000/350/300. 1 * 1 items
Catalog # PERCN8146004
Supplier:  PerkinElmer
Frequently Bought Together