Order Entry
Denmark
ContactUsLinkComponent
PRECISION-DESIGNED AND MANUFACTURED, LARGE-ORIFICE SAMPLER AND SKIMMER CONES PROVIDE SUPERIOR LONG-TERM STABILITY AND RESIST CLOGGIN G, ALLOWING ANALYSIS UNDER BOTH HIGH AND LOW SAMPLE-UPTAKE CONDITIONS. NICKEL IS A VERY RUGGED, LONG-LASTING MATERIAL 1 * 1 items
Catalog # PERCN8161041
Supplier:  PerkinElmer
PRECISION-DESIGNED AND MANUFACTURED, LARGE-ORIFICE SAMPLER AND SKIMMER CONES PROVIDE SUPERIOR LONG-TERM STABILITY AND RESIST CLOGGIN G, ALLOWING ANALYSIS UNDER BOTH HIGH AND LOW SAMPLE-UPTAKE CONDITIONS. NICKEL IS A VERY RUGGED, LONG-LASTING MATERIAL 1 * 1 items
Catalog # PERCN8161041
Supplier:  PerkinElmer
Frequently Bought Together