Order Entry
Denmark
ContactUsLinkComponent
THE ALUMINUM STANDATD EXPANSION 10 MM BY 3 MM D IS AN ALUMINUM REFERENCE MATERIAL FOR THE EXPANSION CALIBRATION OF THE TMA 4000. 1 * 1 items
Catalog # PERCL8041600
Supplier:  PerkinElmer
THE ALUMINUM STANDATD EXPANSION 10 MM BY 3 MM D IS AN ALUMINUM REFERENCE MATERIAL FOR THE EXPANSION CALIBRATION OF THE TMA 4000. 1 * 1 items
Catalog # PERCL8041600
Supplier:  PerkinElmer
Frequently Bought Together