Order Entry
Denmark
ContactUsLinkComponent
Gridding head for protein macroarrays, 96 pin for 96- and 384-well plates, tip diameter 0.15 mm 1 * 1 items
Catalog # MLDVX4227
Supplier:  Molecular Devices
Gridding head for protein macroarrays, 96 pin for 96- and 384-well plates, tip diameter 0.15 mm 1 * 1 items
Catalog # MLDVX4227
Supplier:  Molecular Devices
Frequently Bought Together