Order Entry
Denmark
ContactUsLinkComponent
Picking head for streptomyces and Yeast, 96 pin, tip diameter 1 mm, deep well 1 * 1 items
Catalog # MLDVX4006F
Supplier:  Molecular Devices
Picking head for streptomyces and Yeast, 96 pin, tip diameter 1 mm, deep well 1 * 1 items
Catalog # MLDVX4006F
Supplier:  Molecular Devices
Frequently Bought Together