Order Entry
Denmark
Orders LinkContactUsLinkComponent
Safe aspiration station
Safe aspiration station, EU plug
Katalog # GILSF110744
Leverandør:   GILSON
Safe aspiration station
Katalog # GILSF110744
Leverandør:  GILSON
Safe aspiration station
Safe aspiration station
Safe aspiration station, EU plug
Price
14.972,22 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The Gilson® safe aspiration station is a stand-alone motorised vacuum system for quick and easy aspiration of biological liquid waste disposal. Liquids are quickly and safely aspirated and collected in a 4 L polypropylene bottle. The station offers adjustable aspiration speed and handles, ideal for ELISA tests, cell culture, and any other contaminated or hazardous liquid aspiration and disposal.
  • Safe and contamination-free
  • Adaptable
  • Convenient
The closed, stand-alone unit is designed to prevent contamination hazards. All parts in contact with liquids are fully autoclavable. The station is equipped with a level detection system to prevent liquid overflow and an in-line hydrophobic filter that traps aerosols for safe disposal of contaminated or hazardous liquids.
 : The Gilson® safe aspiration kit contains accessory items that function with the Gilson® safe aspiration stations. Different adapters are provided with the safe aspiration kit and can be fitted to the hand operator. The station also ensures an adjustable aspiration level indicated by a board of LEDs. The choice of the adapter and the easy setting of the vacuum level enables quick and accurate aspiration of liquids from a variety of vessels. Associated with the safe aspiration kit, the safe aspiration station is easy to use on a bench or in laminar flow cabinets. The unit is always ready to use. Simply press the button on the aspiration hand operator to aspirate. The ergonomic hand operator can be stored on the bottle handle. The safe aspiration kit also contains a stand with storage for the adapters and the hand operator to help your workspace remain clean and organised. The Gilson® safe aspiration kit contains one hand operator, one silicon adapter for Pasteur pipettes and silicon grease, silicon grease, one safety stand for the hand operator, one single-channel plastic tip adapter with ejector, one 8-channel plastic tip adapter with ejector, one 8-channel stainless steel needle adapter, four single-channel stainless steel needle adapters (various sizes): 4 cm L×1,5 mm Ø, 4 cm L×3 mm Ø, 12 cm L×1,5 mm Ø, 12 cm L×3 mm Ø.
 : The safe aspiration (accessory) kit is sold separately. For available filters and accessories, contact your Avantor representative.
Specifikationer
  • Description:
    Safe aspiration station, EU plug
  • Seq:
    01
Accessories
Produktinformation
  • Katalog #
    Description
    Tilgængelighed
    Price
  • GILSF1077411
    Bottle, PP, 4 l
    Log ind for tilgængelighed
    1 * 1 Items - 1.487,52 kr
Produktinformation
  • Katalog #
    Description
    Tilgængelighed
    Price
  • GILSF1077418
    Y-tubing connector
    Log ind for tilgængelighed
    1 * 1 Items - 87,28 kr
Anbefalinger