Custom Packed Columns MS-5A 60/80 1.5m 2.0mmID 1/8inOD SILCO HP Std HP Config PreCond NOC KIT-C (former PC25584) 1 * 1 Items
Catalog # RESTPKC31559
Supplier: Restek
Custom Packed Columns MS-5A 60/80 1.5m 2.0mmID 1/8inOD SILCO HP Std HP Config PreCond NOC KIT-C (former PC25584) 1 * 1 Items
Catalog # RESTPKC31559
Supplier: Restek
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK