Order Entry
Denmark
ContactUsLinkComponent
Sulphite Standard Solution 80 mg/l SO2. Duree de conservation : 12 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 100 mL
Catalog # ACSDBP465
Supplier:  CPACHEM
Sulphite Standard Solution 80 mg/l SO2. Duree de conservation : 12 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 100 mL
Catalog # ACSDBP465
Supplier:  CPACHEM
Frequently Bought Together